Assay Details
| Gene Symbol: Rgs9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001165934.1 | NP_001159406.1 | 5 - 6 | 549 | 147 | |
| NM_011268.2 | NP_035398.2 | 5 - 6 | 549 | 147 | ||
| GenBank mRNA | AK085443.1 | - | 5 - 6 | 475 | 147 | |
| AK138159.1 | - | 5 - 6 | 459 | 147 | ||
| AK164961.1 | - | 5 - 6 | 503 | 147 | ||
| BC129899.1 | - | 5 - 6 | 415 | 40111531 | 147 | |
More Information
Set Membership: |
|

Rgs9
Genomic Map