Assay Details
| Gene Symbol: Alox12e | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145684.1 | NP_663717.1 | 10 - 11 | 1472 | 88 | |
| GenBank mRNA | AK138743.1 | - | 10 - 11 | 1426 | 88 | |
| BC013751.1 | - | 10 - 11 | 1452 | 3596809 | 88 | |
| BC051047.1 | - | 10 - 11 | 1456 | 4457881 | 88 | |
| U39200.1 | - | 10 - 11 | 1413 | 88 | ||
| X99252.1 | - | 10 - 11 | 1472 | 88 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Alox12e
Genomic Map