Assay Details
| Gene Symbol: Alox12e | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145684.1 | NP_663717.1 | 5 - 6 | 714 | 112 | |
| GenBank mRNA | AK138743.1 | - | 5 - 6 | 668 | 112 | |
| BC013751.1 | - | 5 - 6 | 694 | 3596809 | 112 | |
| BC051047.1 | - | 5 - 6 | 698 | 4457881 | 112 | |
| U39200.1 | - | 5 - 6 | 655 | 112 | ||
| X99252.1 | - | 5 - 6 | 714 | 112 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Alox12e
Genomic Map