Assay Details
Gene Symbol: Esrp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_176838.2 | NP_789808.1 | 5 - 6 | 858 | 86 | |
GenBank mRNA | AK088755.1 | - | 5 - 6 | 727 | 86 | |
AK168488.1 | - | 5 - 6 | 706 | 86 | ||
BC031444.1 | - | 5 - 6 | 858 | 4222237 | 86 | |
BC096581.1 | - | 5 - 6 | 698 | 86 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
