Assay Details
Gene Symbol: Ranbp10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145824.4 | NP_665823.2 | 5 - 6 | 762 | 74 | |
GenBank mRNA | AK173177.1 | - | 5 - 6 | 575 | 74 | |
AY337314.1 | - | 5 - 6 | 763 | 74 | ||
BC024698.1 | - | 5 - 6 | 607 | 3667000 | 74 | |
EU281316.1 | - | 5 - 6 | 623 | 74 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
