Assay Details
Gene Symbol: Mpp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007863.2 | NP_031889.2 | 2 - 3 | 212 | 142 | |
GenBank mRNA | AF079366.1 | - | 2 - 3 | 163 | 142 | |
AK032270.1 | - | 2 - 3 | 192 | 142 | ||
AK047456.1 | - | 2 - 3 | 192 | 142 | ||
AK163205.1 | - | 2 - 3 | 195 | 142 | ||
BY250841.1 | - | 2 - 3 | 201 | 142 | ||
BY296207.1 | - | 2 - 3 | 214 | 142 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AF079366.1, NM_007863.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
