Assay Details
| Gene Symbol: Fam171a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199200.2 | NP_954670.2 | 6 - 7 | 1046 | 76 | |
| GenBank mRNA | BC025575.1 | - | 6 - 7 | 1022 | 76 | |
| BC064455.1 | - | 3 - 4 | 779 | 6810415 | 76 | |
| CB182176.1 | - | 5 - 6 | 794 | 76 | ||
| CB202938.1 | - | 4 - 5 | 512 | 76 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Fam171a2
Genomic Map