Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0200019513.mm.1 |
NM_001025395.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200019513.mm.1 |
NM_001025395.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200019513.mm.1 |
NM_009271.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200019513.mm.1 |
NM_009271.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200019513.mm.1 |
TC0200002410.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200019518.mm.1 |
NM_001025395.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200019518.mm.1 |
NM_001025395.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200019518.mm.1 |
NM_009271.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200019518.mm.1 |
NM_009271.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200019518.mm.1 |
TC0200002410.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200019511.mm.1 |
NM_001025395.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200019511.mm.1 |
NM_001025395.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200019511.mm.1 |
NM_009271.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200019511.mm.1 |
NM_009271.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200019511.mm.1 |
TC0200002410.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200019514.mm.1 |
NM_001025395.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200019514.mm.1 |
NM_001025395.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200019514.mm.1 |
NM_009271.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200019514.mm.1 |
NM_009271.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200019514.mm.1 |
TC0200002410.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200019515.mm.1 |
NM_001025395.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200019515.mm.1 |
NM_001025395.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200019515.mm.1 |
NM_009271.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200019515.mm.1 |
NM_009271.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200019515.mm.1 |
TC0200002410.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200019507.mm.1 |
NM_001025395.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200019507.mm.1 |
NM_001025395.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200019507.mm.1 |
NM_009271.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200019507.mm.1 |
NM_009271.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200019507.mm.1 |
TC0200002410.mm.1 |
MTA TC |