Assay Details
| Gene Symbol: Src | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025395.2 | NP_001020566.1 | 4 - 5 | 632 | 97 | |
| NM_009271.3 | NP_033297.2 | 4 - 5 | 632 | 97 | ||
| GenBank mRNA | BC039953.1 | - | 4 - 5 | 642 | 4192014 | 97 |
| CX218263.1 | - | 1 - 2 | 131 | 97 | ||
| M17031.1 | - | 1 - 2 | 97 | |||
More Information
Updated Mapping Information
Set Membership: |
|

Src
Genomic Map