Assay Details
| Gene Symbol: Src | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025395.2 | NP_001020566.1 | 9 - 10 | 1244 | 84 | |
| NM_009271.3 | NP_033297.2 | 10 - 11 | 1262 | 84 | ||
| GenBank mRNA | AK146056.1 | - | 1191 | 84 | ||
| AK182969.1 | - | 1 - 2 | 53 | 84 | ||
| AK189521.1 | - | 1 - 2 | 53 | 84 | ||
| BC039953.1 | - | 9 - 10 | 1254 | 4192014 | 84 | |
| M17031.1 | - | 7 - 8 | 878 | 84 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Src
Genomic Map