Assay Details
| Gene Symbol: Tk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021028.3 | NP_066356.3 | 2 - 3 | 196 | 77 | |
| NR_045642.1 | 2 - 3 | 196 | 77 | |||
| GenBank mRNA | AF105217.1 | - | 2 - 3 | 171 | 77 | |
| AJ249341.2 | - | 2 - 3 | 174 | 77 | ||
| AK041651.1 | - | 2 - 3 | 188 | 77 | ||
| AK051286.1 | - | 2 - 3 | 190 | 77 | ||
| AK087557.1 | - | 2 - 3 | 197 | 77 | ||
| AK154160.1 | - | 2 - 3 | 190 | 77 | ||
| BC019982.1 | - | 2 - 3 | 188 | 4159742 | 77 | |
| BY083126.1 | - | 2 - 3 | 191 | 77 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tk2
Genomic Map