Assay Details
| Gene Symbol: Tk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021028.3 | NP_066356.3 | 6 - 7 | 489 | 86 | |
| GenBank mRNA | AF105217.1 | - | 6 - 7 | 464 | 86 | |
| AK041651.1 | - | 6 - 7 | 481 | 86 | ||
| AK087557.1 | - | 6 - 7 | 490 | 86 | ||
| AK154160.1 | - | 6 - 7 | 483 | 86 | ||
| AK187899.1 | - | 2 - 3 | 118 | 86 | ||
| AK196494.1 | - | 2 - 3 | 114 | 86 | ||
| AK202106.1 | - | 2 - 3 | 114 | 86 | ||
| AK216095.1 | - | 2 - 3 | 114 | 86 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tk2
Genomic Map