Assay Details
| Gene Symbol: Ces2g | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_197999.2 | NP_932116.2 | 8 - 9 | 1210 | 135 | |
| GenBank mRNA | AK146261.1 | - | 8 - 9 | 1154 | 135 | |
| BC027185.1 | - | 8 - 9 | 1188 | 4194872 | 135 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ces2g
Genomic Map