Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0800009388.mm.1 |
NM_001164681.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800009388.mm.1 |
NM_001164681.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800009388.mm.1 |
NM_198672.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800009388.mm.1 |
NM_198672.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800009388.mm.1 |
TC0800001200.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800009389.mm.1 |
NM_001164681.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800009389.mm.1 |
NM_001164681.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800009389.mm.1 |
NM_198672.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800009389.mm.1 |
NM_198672.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800009389.mm.1 |
TC0800001200.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800009394.mm.1 |
NM_001164681.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800009394.mm.1 |
NM_001164681.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800009394.mm.1 |
NM_198672.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800009394.mm.1 |
NM_198672.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800009394.mm.1 |
TC0800001200.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800009392.mm.1 |
NM_001164681.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800009392.mm.1 |
NM_001164681.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800009392.mm.1 |
NM_198672.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800009392.mm.1 |
NM_198672.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800009392.mm.1 |
TC0800001200.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800009386.mm.1 |
NM_001164681.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800009386.mm.1 |
NM_001164681.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800009386.mm.1 |
NM_198672.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800009386.mm.1 |
NM_198672.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800009386.mm.1 |
TC0800001200.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800009387.mm.1 |
NM_001164681.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800009387.mm.1 |
NM_001164681.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800009387.mm.1 |
NM_198672.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800009387.mm.1 |
NM_198672.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800009387.mm.1 |
TC0800001200.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800009391.mm.1 |
NM_001164681.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800009391.mm.1 |
NM_001164681.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800009391.mm.1 |
NM_198672.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800009391.mm.1 |
NM_198672.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800009391.mm.1 |
TC0800001200.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800009390.mm.1 |
NM_001164681.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800009390.mm.1 |
NM_001164681.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800009390.mm.1 |
NM_198672.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800009390.mm.1 |
NM_198672.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800009390.mm.1 |
TC0800001200.mm.1 |
MTA TC |