Assay Details
| Gene Symbol: Slc9a5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081332.1 | NP_001074801.1 | 7 - 8 | 2463 | 105 | |
| GenBank mRNA | AK134092.1 | - | 2 - 3 | 777 | 105 | |
| BC057616.1 | - | 3 - 4 | 1218 | 105 | ||
| BC151197.1 | - | 7 - 8 | 1904 | 9088085 | 105 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slc9a5
Genomic Map