Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0800009651.mm.1 |
NM_001033371.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800009651.mm.1 |
NM_001033371.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800009651.mm.1 |
NM_001170789.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800009651.mm.1 |
NM_001170789.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800009651.mm.1 |
TC0800001218.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800009646.mm.1 |
NM_001170788.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800009646.mm.1 |
NM_001170788.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800009646.mm.1 |
TC0800001218.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800009648.mm.1 |
NM_001170788.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800009648.mm.1 |
NM_001170788.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800009648.mm.1 |
TC0800001218.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800009652.mm.1 |
NM_001033371.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800009652.mm.1 |
NM_001033371.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800009652.mm.1 |
NM_001170788.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800009652.mm.1 |
NM_001170788.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800009652.mm.1 |
NM_001170789.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800009652.mm.1 |
NM_001170789.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800009652.mm.1 |
TC0800001218.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800009647.mm.1 |
NM_001170788.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800009647.mm.1 |
NM_001170788.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800009647.mm.1 |
TC0800001218.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800009653.mm.1 |
NM_001033371.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800009653.mm.1 |
NM_001033371.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800009653.mm.1 |
NM_001170788.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800009653.mm.1 |
NM_001170788.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800009653.mm.1 |
NM_001170789.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800009653.mm.1 |
NM_001170789.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800009653.mm.1 |
TC0800001218.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800009649.mm.1 |
NM_001170788.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800009649.mm.1 |
NM_001170788.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800009649.mm.1 |
TC0800001218.mm.1 |
MTA TC |