Assay Details
Gene Symbol: Zdhhc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175160.3 | NP_780369.1 | 4 - 5 | 419 | 66 | |
GenBank mRNA | AK038593.1 | - | 4 - 5 | 421 | 66 | |
BC119378.1 | - | 1 - 2 | 158 | 8734127 | 66 | |
BC144878.1 | - | 1 - 2 | 158 | 9053426 | 66 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AK038593.1, BC119378.1, BC144878.1, NM_175160.3 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
