Assay Details
Gene Symbol: Zdhhc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175160.3 | NP_780369.1 | 6 - 7 | 846 | 57 | |
GenBank mRNA | AK014489.1 | - | 1 - 2 | 1714 | 57 | |
AK038593.1 | - | 6 - 7 | 848 | 57 | ||
BC026570.1 | - | 2 - 3 | 427 | 4984252 | 57 | |
BC119378.1 | - | 3 - 4 | 585 | 8734127 | 57 | |
BC144878.1 | - | 3 - 4 | 585 | 9053426 | 57 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
