Assay Details
Gene Symbol: Cwf19l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027545.2 | NP_081821.1 | 11 - 12 | 1731 | 82 | |
GenBank mRNA | AK045423.1 | - | 11 - 12 | 1732 | 82 | |
AK083386.1 | - | 11 - 12 | 1744 | 82 | ||
AK167727.1 | - | 4 - 5 | 454 | 82 | ||
BC043480.1 | - | 11 - 12 | 1698 | 82 | ||
BC125483.1 | - | 11 - 12 | 1768 | 40129998 | 82 | |
BC125485.1 | - | 11 - 12 | 1768 | 40130000 | 82 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
