Assay Details
| Gene Symbol: 1700030J22Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027103.2 | NP_081379.1 | 2 - 3 | 410 | 71 | |
| GenBank mRNA | AK006553.1 | - | 2 - 3 | 415 | 71 | |
| AK030075.1 | - | 2 - 3 | 411 | 71 | ||
| AK031529.1 | - | 2 - 3 | 295 | 71 | ||
| BC100501.1 | - | 2 - 3 | 430 | 30917058 | 71 | |
| BC137750.1 | - | 2 - 3 | 348 | 8860769 | 71 | |
| BC137751.1 | - | 2 - 3 | 348 | 8860770 | 71 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26200046 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26200045 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26200041 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26200043 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26200039 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26200040 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P670735 | BC100501.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P670735 | BC137750.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P670735 | BC137751.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P670735 | AK030075.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P670735 | AK031529.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P670735 | BC100501.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P670735 | BC137750.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P670735 | BC137751.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P670735 | AK030075.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P670735 | AK031529.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 404438 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 881686 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 217506 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 724791 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 907927 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 31572 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P136444 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P148997 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P670735 | BC100501.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P670735 | BC137750.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P670735 | BC137751.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P670735 | AK030075.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P670735 | AK031529.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P670735 | BC100501.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P670735 | BC137750.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P670735 | BC137751.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P670735 | AK030075.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P670735 | AK031529.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26200059 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26200060 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26200050 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26200053 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26200044 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26200052 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26200051 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26200039 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26200031 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26200056 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26200049 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26200046 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26200048 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26200035 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26200034 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26200055 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26200041 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06655560 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P148997 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P136444 | - | - |
| Whole Mouse Genome G4122A | A_52_P670735 | BC100501.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P670735 | BC137750.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P670735 | BC137751.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P670735 | AK030075.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P670735 | AK031529.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P670735 | BC100501.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P670735 | BC137750.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P670735 | BC137751.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P670735 | AK030075.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P670735 | AK031529.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P148997 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P136444 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07154535 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07154539 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07154531 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06744107 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07154538 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06744104 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06744106 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26200041 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26200043 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26200040 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26200039 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26200046 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26200045 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26200057 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26200048 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26200026 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26199985 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26199981 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26200050 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26200052 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26199936 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26200085 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26199934 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26199970 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26200029 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26199969 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26200003 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26200060 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26200007 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26200046 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26199983 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26200022 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26200036 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26199996 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26200059 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26199998 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26200024 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26200004 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26200082 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26200090 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26200083 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26200015 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26199960 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26200000 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26200044 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26199987 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26199939 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26200028 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26200032 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26199991 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26200010 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26200011 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26200009 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26199984 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26199978 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26200039 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26200041 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26199938 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26200001 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26200053 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26200086 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26200093 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26199941 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26199963 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26200027 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26199993 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26200092 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26199982 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26200021 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26200017 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26200032 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26200053 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26200026 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26200035 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26200022 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26200002 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06935289 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26200005 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26200011 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26200039 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26200021 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26200052 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07148236 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26200055 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26200031 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26200010 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26199998 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26200003 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26200015 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26200057 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07059516 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26200028 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26200041 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07059514 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26200059 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26200000 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26200044 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07059517 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06854816 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26200046 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26200004 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26200001 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07059515 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26200017 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26200050 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26200051 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26200048 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26200056 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07005126 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26200029 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26200058 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07154533 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07005130 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26199996 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26200020 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10582019 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10582016 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10582018 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10582017 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10582015 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1430565_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10582014 | - | - |
| Mouse Genome 430 2.0 Array | 1430565_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1430565_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 69528_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 69528_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0800023095.mm.1 | NM_027103.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800023095.mm.1 | NM_027103.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800023095.mm.1 | TC0800003007.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0800023096.mm.1 | NM_027103.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800023096.mm.1 | NM_027103.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800023096.mm.1 | TC0800003007.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0800023094.mm.1 | NM_027103.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800023094.mm.1 | NM_027103.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800023094.mm.1 | TC0800003007.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1430565_PM_at | - | - |
| Mouse Expression Set 430 | 1430565_at | - | - |
| Murine Genome U74v2 Set | 129740_PM_at | - | - |
| Murine Genome U74v2 Set | 139155_PM_at | - | - |
| Murine Genome U74v2 Set | 129740_at | - | - |
| Murine Genome U74v2 Set | 139155_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1430565_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1430565_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 311131 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 3360504 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1231468 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1231468 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1231468 | - | - |

1700030J22Rik
Genomic Map