Assay Details
Gene Symbol: Cdyl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_029441.3 | NP_083717.1 | 1 - 2 | 195 | 91 | |
GenBank mRNA | AK142084.1 | - | 1 - 2 | 85 | 91 | |
BC058956.1 | - | 1 - 2 | 195 | 5690642 | 91 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AK142084.1, BC058956.1, NM_029441.3 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
