Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0200018021.mm.1 |
NM_001081044.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200018021.mm.1 |
NM_001081044.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200018021.mm.1 |
TC0200002303.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200018022.mm.1 |
NM_001081044.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200018022.mm.1 |
NM_001081044.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200018022.mm.1 |
TC0200002303.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200018024.mm.1 |
NM_001081044.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200018024.mm.1 |
NM_001081044.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200018024.mm.1 |
TC0200002303.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200018023.mm.1 |
NM_001081044.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200018023.mm.1 |
NM_001081044.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200018023.mm.1 |
TC0200002303.mm.1 |
MTA TC |