Assay Details
Gene Symbol: Mylk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081044.2 | NP_001074513.2 | 6 - 7 | 1336 | 89 | |
GenBank mRNA | AK079396.1 | - | 6 - 7 | 1343 | 89 | |
BC158041.1 | - | 6 - 7 | 1167 | 9088157 | 89 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
