Assay Details
| Gene Symbol: Cdk5rap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025876.2 | NP_080152.1 | 11 - 12 | 1454 | 67 | |
| GenBank mRNA | AK010081.1 | - | 11 - 12 | 1454 | 67 | |
| AK010097.1 | - | 11 - 12 | 1454 | 67 | ||
| AK088517.1 | - | 11 - 12 | 1463 | 67 | ||
| BC025132.1 | - | 10 - 11 | 1419 | 3666341 | 67 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cdk5rap1
Genomic Map