Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0800011327.mm.1 |
NM_001163028.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800011327.mm.1 |
NM_001163028.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800011327.mm.1 |
NM_021486.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800011327.mm.1 |
NM_021486.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800011327.mm.1 |
TC0800001396.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800011328.mm.1 |
NM_001163028.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800011328.mm.1 |
NM_001163028.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800011328.mm.1 |
NM_021486.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800011328.mm.1 |
NM_021486.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800011328.mm.1 |
TC0800001396.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800011329.mm.1 |
NM_001163028.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800011329.mm.1 |
NM_001163028.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800011329.mm.1 |
NM_021486.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800011329.mm.1 |
NM_021486.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800011329.mm.1 |
TC0800001396.mm.1 |
MTA TC |