Assay Details
| Gene Symbol: Dlgap4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001277186.1 | NP_001264115.1 | 6 - 7 | 1833 | 60 | |
| NM_146128.6 | NP_666240.4 | 6 - 7 | 1833 | 60 | ||
| GenBank mRNA | AK122408.1 | - | 6 - 7 | 1833 | 60 | |
| AY243849.2 | - | 4 - 5 | 1413 | 60 | ||
| BC141110.1 | - | 4 - 5 | 9055722 | 60 | ||
| BC145394.1 | - | 4 - 5 | 1470 | 9053952 | 60 | |
More Information
Updated Mapping Information
Set Membership: |
|

Dlgap4
Genomic Map