Assay Details
| Gene Symbol: Ace3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001101453.2 | NP_001094923.2 | 11 - 12 | 1548 | 87 | |
| GenBank mRNA | AB531024.1 | - | 11 - 12 | 1548 | 87 | |
| CF105940.1 | - | 3 - 4 | 278 | 87 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ace3
Genomic Map