Assay Details
| Gene Symbol: Ace3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001101453.2 | NP_001094923.2 | 13 - 14 | 1856 | 78 | |
| GenBank mRNA | AB531024.1 | - | 13 - 14 | 1856 | 78 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ace3
Genomic Map