Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0800007190.mm.1 |
NM_001164559.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800007190.mm.1 |
NM_001164559.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800007190.mm.1 |
NM_001164560.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800007190.mm.1 |
NM_001164560.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800007190.mm.1 |
NM_198020.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800007190.mm.1 |
NM_198020.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800007190.mm.1 |
TC0800003233.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800049128.mm.1 |
NM_001164559.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800049128.mm.1 |
NM_001164559.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800049128.mm.1 |
NM_001164560.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800049128.mm.1 |
NM_001164560.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800049128.mm.1 |
NM_198020.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800049128.mm.1 |
NM_198020.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800049128.mm.1 |
TC0800003233.mm.1 |
MTA TC |