Assay Details
| Gene Symbol: Efcab3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081046.1 | NP_001074515.1 | 4 - 5 | 592 | 145 | |
| GenBank mRNA | AK014861.1 | - | 4 - 5 | 586 | 145 | |
| AK162207.1 | - | 13 - 14 | 1077 | 145 | ||
| BC050801.1 | - | 4 - 5 | 590 | 6770929 | 145 | |
More Information
Updated Mapping Information
Set Membership: |
|

Efcab3
Genomic Map