Assay Details
| Gene Symbol: Efcab3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081046.1 | NP_001074515.1 | 7 - 8 | 970 | 126 | |
| GenBank mRNA | BC050801.1 | - | 7 - 8 | 968 | 6770929 | 126 |
More Information
Updated Mapping Information
Set Membership: |
|

Efcab3
Genomic Map