Assay Details
| Gene Symbol: Gcdh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044744.1 | NP_001038209.2 | 1 - 2 | 181 | 81 | |
| GenBank mRNA | BY301880.1 | - | 1 - 2 | 182 | 81 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001044744.1 transcripts.
Set Membership: |
|

Gcdh
Genomic Map