Assay Details
| Gene Symbol: Serinc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_012032.4 | NP_036162.3 | 3 - 4 | 506 | 133 | |
| GenBank mRNA | AB029499.1 | - | 3 - 4 | 415 | 133 | |
| AF181684.1 | - | 3 - 4 | 434 | 133 | ||
| AK145604.1 | - | 3 - 4 | 454 | 133 | ||
| AK150106.1 | - | 3 - 4 | 452 | 133 | ||
| AK152720.1 | - | 3 - 4 | 452 | 133 | ||
| AK155921.1 | - | 3 - 4 | 452 | 133 | ||
| AK159347.1 | - | 3 - 4 | 452 | 133 | ||
| AK159488.1 | - | 3 - 4 | 452 | 133 | ||
| AK159819.1 | - | 3 - 4 | 452 | 133 | ||
| AK160005.1 | - | 3 - 4 | 452 | 133 | ||
| AK160055.1 | - | 3 - 4 | 452 | 133 | ||
| AK161981.1 | - | 3 - 4 | 452 | 133 | ||
| AK164807.1 | - | 3 - 4 | 452 | 133 | ||
| AK166924.1 | - | 3 - 4 | 452 | 133 | ||
| AK170350.1 | - | 3 - 4 | 445 | 133 | ||
| BC011295.1 | - | 3 - 4 | 462 | 3487943 | 133 | |
| BC022901.1 | - | 3 - 4 | 443 | 5068196 | 133 | |
| BC029026.1 | - | 3 - 4 | 471 | 5039323 | 133 | |
| BC029980.1 | - | 3 - 4 | 475 | 133 | ||
| BC058550.1 | - | 179 | 133 | |||
| L29441.1 | - | 2 - 3 | 231 | 133 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Serinc3
Genomic Map