Assay Details
| Gene Symbol: Serinc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_012032.4 | NP_036162.3 | 4 - 5 | 575 | 90 | |
| GenBank mRNA | AB029499.1 | - | 4 - 5 | 484 | 90 | |
| AF181684.1 | - | 4 - 5 | 503 | 90 | ||
| AK031101.1 | - | 4 - 5 | 564 | 90 | ||
| AK128955.1 | - | 2 - 3 | 112 | 90 | ||
| AK145604.1 | - | 4 - 5 | 523 | 90 | ||
| AK150106.1 | - | 4 - 5 | 521 | 90 | ||
| AK152720.1 | - | 4 - 5 | 521 | 90 | ||
| AK155921.1 | - | 4 - 5 | 521 | 90 | ||
| AK159347.1 | - | 4 - 5 | 521 | 90 | ||
| AK159488.1 | - | 4 - 5 | 521 | 90 | ||
| AK159819.1 | - | 4 - 5 | 521 | 90 | ||
| AK160005.1 | - | 4 - 5 | 521 | 90 | ||
| AK160055.1 | - | 4 - 5 | 521 | 90 | ||
| AK161981.1 | - | 4 - 5 | 521 | 90 | ||
| AK164807.1 | - | 4 - 5 | 521 | 90 | ||
| AK166924.1 | - | 4 - 5 | 521 | 90 | ||
| AK170350.1 | - | 4 - 5 | 514 | 90 | ||
| BC029980.1 | - | 4 - 5 | 544 | 90 | ||
| BC058550.1 | - | 248 | 90 | |||
| L29441.1 | - | 3 - 4 | 300 | 90 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Serinc3
Genomic Map