Assay Details
| Gene Symbol: Pabpc1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001114079.2 | NP_001107551.1 | 6 - 7 | 1136 | 65 | |
| GenBank mRNA | BC158030.1 | - | 6 - 7 | 1136 | 9007333 | 65 |
| EU669908.1 | - | 6 - 7 | 879 | 65 | ||
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Transcript Location | Amplicon Length | |
|---|---|---|---|---|---|---|---|
| GenBankmRNA | DV656812.1 | - | 250 | Chr. - - [-] on Build | 65 | ||
More Information
Set Membership: |
|

Pabpc1l
Genomic Map