Assay Details
| Gene Symbol: Rbpjl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009036.1 | NP_033062.1 | 9 - 10 | 1118 | 82 | |
| GenBank mRNA | AK052449.1 | - | 7 - 8 | 4229 | 82 | |
| AK131814.1 | - | 9 - 10 | 1099 | 82 | ||
| BC152311.1 | - | 9 - 10 | 1112 | 40090714 | 82 | |
| Y10926.1 | - | 9 - 10 | 1118 | 82 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rbpjl
Genomic Map