Assay Details
| Gene Symbol: Cherp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_138585.3 | NP_613051.3 | 7 - 8 | 1024 | 59 | |
| GenBank mRNA | AF536541.1 | - | 7 - 8 | 926 | 59 | |
| AK049765.1 | - | 7 - 8 | 934 | 59 | ||
| AK144328.1 | - | 7 - 8 | 942 | 59 | ||
| AK220122.1 | - | 1 - 2 | 58 | 59 | ||
| BC032191.1 | - | 7 - 8 | 948 | 5322208 | 59 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cherp
Genomic Map