Assay Details
| Gene Symbol: Spns3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC150778.1 | - | 1 - 2 | 229 | 9087689 | 101 |
More Information
Updated Mapping Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Spns3| Gene Symbol: Spns3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC150778.1 | - | 1 - 2 | 229 | 9087689 | 101 |
Set Membership: |
|