Assay Details
| Gene Symbol: Shpk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029031.3 | NP_083307.1 | 2 - 3 | 337 | 73 | |
| GenBank mRNA | AK015273.1 | - | 2 - 3 | 360 | 73 | |
| BC054721.1 | - | 2 - 3 | 337 | 5694588 | 73 | |
| BC056433.1 | - | 2 - 3 | 337 | 5694588 | 73 | |
More Information
Updated Mapping Information
Set Membership: |
|

Shpk
Genomic Map