Assay Details
| Gene Symbol: Caskin2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_080643.2 | NP_542374.2 | 11 - 12 | 1671 | 124 | |
| GenBank mRNA | AK129297.1 | - | 11 - 12 | 1709 | 124 | |
| BC053083.1 | - | 11 - 12 | 1671 | 5704642 | 124 | |
More Information
Updated Mapping Information
Set Membership: |
|

Caskin2
Genomic Map