Assay Details
| Gene Symbol: Nat9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025400.3 | NP_079676.1 | 1 - 2 | 200 | 90 | |
| GenBank mRNA | AK003948.1 | - | 1 - 2 | 154 | 90 | |
| AK082469.1 | - | 1 - 2 | 140 | 90 | ||
| AK148048.1 | - | 1 - 2 | 232 | 90 | ||
| AK154593.1 | - | 1 - 2 | 160 | 90 | ||
| BC011315.1 | - | 1 - 2 | 200 | 4013700 | 90 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK003948.1,AK082469.1,AK148048.1,BC011315.1,NM_025400.3 transcripts.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Nat9
Genomic Map