Assay Details
Gene Symbol: Recql5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_130454.2 | NP_569721.1 | 5 - 6 | 1559 | 82 | |
GenBank mRNA | AB059999.1 | - | 5 - 6 | 1073 | 82 | |
AK029060.1 | - | 5 - 6 | 1213 | 82 | ||
AK050965.1 | - | 3 - 4 | 288 | 82 | ||
AK083513.1 | - | 5 - 6 | 1555 | 82 | ||
BC023769.1 | - | 4 - 5 | 851 | 82 | ||
BC053086.1 | - | 5 - 6 | 1812 | 82 | ||
BC059000.1 | - | 5 - 6 | 1533 | 82 | ||
BC060070.1 | - | 5 - 6 | 1812 | 82 | ||
BC066176.1 | - | 5 - 6 | 1282 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
