| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0800019073.mm.1 |
NM_138944.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800019073.mm.1 |
NM_138944.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800019073.mm.1 |
TC0800002508.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800019074.mm.1 |
NM_138944.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800019074.mm.1 |
NM_138944.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800019074.mm.1 |
TC0800002508.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800019075.mm.1 |
NM_138944.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800019075.mm.1 |
NM_138944.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800019075.mm.1 |
TC0800002508.mm.1 |
MTA TC |