Assay Details
Gene Symbol: Qrich2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033267.2 | NP_001028439.1 | 3 - 4 | 656 | 115 | |
GenBank mRNA | AK131945.1 | - | 3 - 4 | 659 | 115 | |
BC150976.1 | - | 3 - 4 | 509 | 9087889 | 115 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
