Assay Details
Gene Symbol: Mgat5b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172948.3 | NP_766536.2 | 6 - 7 | 1302 | 91 | |
GenBank mRNA | AB116028.1 | - | 6 - 7 | 689 | 91 | |
AB119127.1 | - | 6 - 7 | 768 | 91 | ||
BC094604.1 | - | 6 - 7 | 1302 | 6822690 | 91 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
