Assay Details
Gene Symbol: Mgat5b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172948.3 | NP_766536.2 | 8 - 9 | 1635 | 82 | |
GenBank mRNA | AB116028.1 | - | 8 - 9 | 1022 | 82 | |
AB119127.1 | - | 8 - 9 | 1101 | 82 | ||
BC063084.1 | - | 7 - 8 | 1538 | 82 | ||
BC094604.1 | - | 8 - 9 | 1635 | 6822690 | 82 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
