Assay Details
| Gene Symbol: Acox1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001271898.1 | NP_001258827.1 | 2 - 3 | 613 | 72 | |
| GenBank mRNA | AB034914.1 | - | 2 - 3 | 302 | 72 | |
| AF006688.1 | - | 2 - 3 | 352 | 72 | ||
| AK128941.1 | - | 2 - 3 | 315 | 72 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Acox1
Genomic Map