Assay Details
Gene Symbol: Ten1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027107.1 | NP_081383.1 | 3 - 4 | 423 | 76 | |
GenBank mRNA | AK009156.1 | - | 3 - 4 | 390 | 76 | |
AK146257.1 | - | 3 - 4 | 413 | 76 | ||
BC027639.1 | - | 3 - 4 | 368 | 1069092 | 76 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P431554 | AK146257.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P431554 | BC027639.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P431554 | AK009156.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P431554 | NM_027107.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P431554 | AK146257.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P431554 | BC027639.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P431554 | AK009156.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P431554 | NM_027107.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 206156 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 822724 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 691592 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 168423 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 946822 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 508552 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 639269 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 682143 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 76843 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 4108 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 122836 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P143143 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P161158 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P431554 | AK146257.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P431554 | BC027639.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P431554 | AK009156.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P431554 | NM_027107.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P431554 | AK146257.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P431554 | BC027639.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P431554 | AK009156.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P431554 | NM_027107.1 | mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07504804 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P161158 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P143143 | - | - |
Whole Mouse Genome G4122A | A_51_P431554 | AK146257.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P431554 | BC027639.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P431554 | AK009156.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P431554 | NM_027107.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P431554 | AK146257.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P431554 | BC027639.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P431554 | AK009156.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P431554 | NM_027107.1 | mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P161158 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P143143 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28157991 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09399095 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28157993 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28157996 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10382778 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10382775 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10382776 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10382777 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1427895_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10382774 | - | - |
Mouse Genome 430 2.0 Array | 1427895_at | - | - |
Mouse Genome 430 2.0 Array | 1427895_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 69535_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 69535_PM_at | - | - |
Affymetrix Mouse Clariom S Array | PSR1100018289.mm.1 | NM_027107.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100018289.mm.1 | NM_027107.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100018289.mm.1 | TC1100001924.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1100018288.mm.1 | NM_027107.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100018288.mm.1 | NM_027107.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100018288.mm.1 | TC1100001924.mm.1 | MTA TC |
Mouse Expression Set 430 | 1427895_PM_at | - | - |
Mouse Expression Set 430 | 1427895_at | - | - |
Murine Genome U74v2 Set | 99157_PM_at | - | - |
Murine Genome U74v2 Set | 99157_at | - | - |
Mouse Genome 430A 2.0 Array | 1427895_at | - | - |
Mouse Genome 430A 2.0 Array | 1427895_PM_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1427895_PM_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1427895_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 416004 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2693591 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2693591 | - | - |
