Assay Details
| Gene Symbol: Nanos3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_194059.2 | NP_918948.1 | 1 | 266 | 65 | |
| GenBank mRNA | AB095973.1 | - | 1 | 266 | 65 | |
| BC119513.1 | - | 1 | 266 | 40055381 | 65 | |
| BC119514.1 | - | 1 | 266 | 40055383 | 65 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Nanos3
Genomic Map