Assay Details
| Gene Symbol: Olfr393 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_147008.2 | NP_667219.1 | 1 | 740 | 136 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P197274 | NM_147008.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P197274 | NM_147008.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P197274 | NM_147008.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P197274 | NM_147008.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P197274 | NM_147008.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P197274 | NM_147008.2 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27918293 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27918296 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27918315 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27918268 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27918300 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27918299 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27918267 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27918274 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27918280 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27918275 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27918317 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27918292 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27918304 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27918291 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27918303 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27918307 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27918316 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27918278 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27918284 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27918279 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27918309 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27918311 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27918302 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27918271 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27918306 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27918288 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27918290 | - | - |
| Whole Mouse Genome G4122A | A_51_P197274 | NM_147008.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P197274 | NM_147008.2 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P130373 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08993986 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09275544 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08993985 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09275538 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08993990 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27918262 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27918267 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27918303 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27918317 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27918296 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27918290 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27918274 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27918248 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27918291 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27918292 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27918234 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27918280 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27918246 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27918302 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27918268 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27918312 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27918278 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27918231 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27918240 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27918271 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27918258 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27918300 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27918309 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27918252 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27918304 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27918275 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27918238 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27918288 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27918249 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27918232 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27918316 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27918269 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27918311 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27918307 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27918279 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27918251 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27918284 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27918255 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27918230 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27918299 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27918236 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27918271 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27918309 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27918291 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27918288 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27918249 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27918266 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27918275 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27918302 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27918317 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27918290 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27918311 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27918255 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27918299 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27918267 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27918312 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27918268 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27918307 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27918296 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27918283 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27918280 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27918247 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27918274 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27918248 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27918304 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27918284 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27918261 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27918316 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27918279 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27918300 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27918303 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27918260 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09463787 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27918258 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27918264 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27918292 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27918252 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27918278 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27918256 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10388291 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10388290 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 473829 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 102340685 | - | - |
| MouseWG-6 v2 Expression BeadChip | 510242 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 102340685 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 510242 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0000510242 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1237487 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1237487 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1227271 | - | - |
| mouse-6 v1.1 (Illumina) | GI_22128920-S | - | - |
| mouse-6 v1.1 (Illumina) | scl13061.1.1_51-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 510242 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 102340685 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl13061.1.1_51-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 102340685 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 510242 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1237487 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1237487 | - | - |

Olfr393
Genomic Map